missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA C-gamma Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93404-0.1ml
This item is not returnable.
View return policy
Description
PKA C-gamma Polyclonal antibody specifically detects PKA C-gamma in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PKA C-gamma | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| cAMP-dependent protein kinase catalytic subunit gamma, EC 2.7.11, EC 2.7.11.11, KAPG, PKA C-gamma, PKACg, protein kinase, cAMP-dependent, catalytic, gamma, serine(threonine) protein kinase | |
| A synthetic peptide corresponding to a sequence within amino acids 1-351 of human PKA C-gamma (NP_002723.2). MGNAPAKKDTEQEESVNEFLAKARGDFLYRWGNPAQNTASSDQFERLRTLGMGSFGRVMLVRHQETGGHYAMKILNKQKVVKMKQVEHILNEKRILQAIDFPFLVKLQFSFKDNSYLYLVMEYVPGGEMFSRLQRVGRFSEPHACFYAAQVVLAVQYLHSLDLIHRDLKPENLLIDQQGYLQVTDFGFAKRVKGRTWTLCGTPEYLAPEIILSKGYNKAVDWWALGVLIYEMAVGFPPFYADQPIQIYEKIVSGRVRFPSKLSSDLKHLLRSLLQVDLTKRFGNLRNGVGDIKNHKWFATTSWIAIYEKKVEAPFIPKYTGPGDASNFDDYEEEELRISINEKCAKEFSEF | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 5568 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction