missing translation for 'onlineSavingsMsg'
Learn More

PKA C beta Antibody (2G8-1D12), Novus Biologicals™

Product Code. 18377288 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377288 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377288 Supplier Novus Biologicals Supplier No. H00005567M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKA C beta Monoclonal antibody specifically detects PKA C beta in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKA C beta
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2G8-1D12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH16285
Gene Alias cAMP-dependent protein kinase catalytic beta subunit isoform 4ab, cAMP-dependent protein kinase catalytic subunit beta, DKFZp781I2452, EC 2.7.11, EC 2.7.11.11, MGC41879, MGC9320, PKA C-beta, PKACb, protein kinase A catalytic subunit beta, protein kinase, cAMP-dependent, catalytic, beta
Host Species Mouse
Immunogen PRKACB (AAH16285, 1 a.a. ~ 257 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGNAATAKKGSEVESVKEFLAKAKEDFLKKWENPTQNNAGLEDFERKKTLGTGSFGRVMLVKHKATEQYYAMKILDKQKVVKLKQIEHTLNEKRILQAVNFPFLVRLEYAFKDNSNLYMVMEYVPGGEMFSHLRRIGRFSEPHARFYAAQIVLTFEYLHSLDLIYRDLKPENLLIDHQGYIQVTDFGFAKRVKGRTWTLCGTPEYLAPEIILSKGYNKAVDWWALGVLIYEMAAGYPPFFADQPIQIYEKIVSGKNF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5567
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.