missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKA C alpha/beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115763
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PKA alpha/beta CAT.
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. PRKACB is a member of the Ser/Thr protein kinase family and is a catalytic subunit of AMPK.
Specifications
| PKA C alpha/beta | |
| Polyclonal | |
| Unconjugated | |
| PRKACA | |
| cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit beta; cAMP-dependent protein kinase catalytic subunit C alpha; Cs-PKA; PKA C-alpha; PKA C-beta; Pkaca; Pkacb; PKCA1; PKCD; PPNAD4; Prkaca; Prkacb; protein kinase A; protein kinase A catalytic subunit; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit alpha; protein kinase cAMP-activated catalytic subunit beta; protein kinase, cAMP dependent, catalytic, alpha; protein kinase, cAMP dependent, catalytic, beta; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; protein kinase, cAMP-dependent, catalytic, beta; sperm cAMP-dependent protein kinase catalytic subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18747, 18749, 25636, 293508, 5566, 5567 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P05132, P17612, P22694, P27791, P68181, P68182 | |
| PRKACA, PRKACB | |
| A synthesized peptide derived from human PRKACA(Accession P17612), corresponding to amino acid residues M1-I47. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction