missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA C alpha Antibody (7H3A4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £435.00
Specifications
| Antigen | PKA C alpha |
|---|---|
| Clone | 7H3A4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18192949
|
Novus Biologicals
NBP2-37246 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667395
|
Novus Biologicals
NBP2-37246-0.025ml |
0.025 mL |
£187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKA C alpha Monoclonal specifically detects PKA C alpha in Human samples. It is validated for Western Blot, ELISA.Specifications
| PKA C alpha | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| cAMP-dependent protein kinase catalytic subunit alpha, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, PKA C-alpha, PKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha | |
| PRKACA | |
| IgG1 | |
| Affinity Purified |
| 7H3A4 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 5566 | |
| Purified recombinant fragment of human PRKACA (AA: 1-120) expressed in E. Coli. | |
| Primary | |
| 40.6 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title