missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKA C alpha Antibody (6D2.1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62195B
This item is not returnable.
View return policy
Description
PKA C alpha Monoclonal specifically detects PKA C alpha in Human samples. It is validated for Western Blot.
Specifications
| PKA C alpha | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PRKACA | |
| Peptide corresponding to amino acids ESPAQNTAHLDQFERIK of human proteinkinase A c alpha (PKAc alpha). | |
| 0.1 mL | |
| Primary | |
| The antibody C-50 reacts with Cytokeratin peptides 5 and 18 (58 and 45 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues. | |
| Store at 4°C in the dark. |
| Western Blot | |
| 6D2.1 | |
| Western Blot | |
| cAMP-dependent protein kinase catalytic subunit alpha, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, PKA C-alpha, PKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha | |
| Mouse | |
| Protein A purified | |
| Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 5566 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction