Learn More
PKA alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA130236
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-30236 detects PKA from mouse, human, rat, bovine samples. PA1-30236 has been successfully used in immunoprecipitation, Western blot applications. The PA1-30236 immunogen is: Synthetic peptide (residues 7-21) based on the human 40 kDa alpha isoform of the catalytic subunit of PKA.
Protein Kinase A (PKA) is the kinase that mediates the majority of the actions of the intracellular second messenger cAMP in diverse tissues and phyla. PKA regulates a number of cellular functions including gene expression, carbohydrate metabolism, Ca2+Spécification
PKA alpha | |
Polyclonal | |
Unconjugated | |
P00517, P17612, P05132, P27791 | |
PRKACA | |
Synthetic peptide (residues 7-21) based on the human 40 kDa alpha isoform of the catalytic subunit of PKA. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PRKACA | |
PKA, cAMP-Dependent Protein Kinase, Catalytic Subunit PKA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18747, 25636, 282322, 5566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |