missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PJA2/Praja2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | PJA2/Praja2 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18396876
|
Novus Biologicals
NBP3-03730-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18353407
|
Novus Biologicals
NBP3-03730-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PJA2/Praja2 Polyclonal specifically detects PJA2/Praja2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PJA2/Praja2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| E3 ubiquitin-protein ligase Praja-2, EC 6.3.2, EC 6.3.2.-, KIAA0438praja2, Neurodap1, praja ring finger 2, Praja2, RING finger protein 131praja 2, RING-H2 motif containing, RNF131 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 439-708 of human PJA2/Praja2 (NP_055634.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS with 50% glycerol, pH7.3. | |
| 9867 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title