missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PJA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572845
This item is not returnable.
View return policy
Description
PJA Polyclonal Antibody for Western Blot, IHC (P), ELISA
Has E2-dependent E3 ubiquitin-protein ligase activity. Ubiquitinates MAGED1 antigen leading to its subsequent degradation by proteasome. May be involved in protein sorting.
Specifications
| PJA | |
| Polyclonal | |
| Unconjugated | |
| PJA1 | |
| E3 ubiquitin-protein ligase Praja-1; Pja1; praja 1; praja ring finger 1, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 1; Praja1; praja-1; praja1, RING-H2 motif containing; RING finger protein 70; RING-type E3 ubiquitin transferase Praja-1; RNF70 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18744, 64219, 683077 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O55176, Q8NG27 | |
| PJA1 | |
| PJA1 antibody for a 19 amino acid peptide near the amino terminus of human PJA1. The immunogen is within amino acids 50 - 100 of PJA1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction