missing translation for 'onlineSavingsMsg'
Learn More

PJ34 hydrochloride, MedChemExpress

Product Code. 16496276
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496276 100 mg 100mg
16485808 10 mg 10mg
16421967 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496276 Supplier Medchem Express Supplier No. HY13688
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PJ34 hydrochloride is an inhibitor of PARP1/2 with IC50 of 110 nM and 86 nM, respectively.

PJ34 hydrochloride is an inhibitor of PARP1/2 with IC50 of 110 nM and 86 nM, respectively.

Specifications

Chemical Name or Material PJ34 hydrochloride
Physical Form Solid
CAS 344458-15-7
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H18ClN3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : 50 mg/mL (150.69 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (37.67 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC(C=C1C2=C3C=CC=C2)=CC=C1NC3=O)CN(C)C.Cl
Molecular Weight (g/mol) 331.8
Formula Weight 331.8
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.