missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PJ 34 hydrochloride

Product Code. 18054432
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054432 50 mg 50mg
18025812 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054432 Supplier R&D Systems™ Supplier No. 3255/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent PARP inhibitor

Potent inhibitor of poly(ADP-ribose) polymerase (PARP) (EC50 = 20 nM). ∽1000-fold more potent than 3-Aminobenzamide (Cat. No. 0788). Protects primary neuronal cells from oxygen-glucose deprivation in vitro and reduces infarct size following focal cerebral ischemia in vivo. Displays protective effects against cisplatin -induced kidney injury. Also displays activity at Pim-1 and Pim-2 kinases at higher concentrations (IC50 values are 3.7 and 16 μM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344458-15-7
Molecular Formula C17H18ClN3O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 331.8
Purity >98%
Molecular Weight (g/mol) 331.8
Chemical Name or Material PJ 34 hydrochloride
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.