missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PIWIL4 Polyclonal Antibody

PIWIL4 Polyclonal Antibody
Brand: Thermo Scientific PA557731
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PIWIL4 Polyclonal Antibody for IHC
Tekniske data
| PIWIL4 | |
| Polyclonal | |
| Unconjugated | |
| PIWIL4 | |
| 9230101H05Rik; HIWI2; mAgo5; Miwi2; PIWI; piwi like RNA-mediated gene silencing 4; Piwil4; piwi-like 4; piwi-like homolog 4; piwi-like protein 4; piwi-like RNA-mediated gene silencing 4; testis tissue sperm-binding protein Li 85P | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 143689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z3Z4 | |
| PIWIL4 | |
| Recombinant protein corresponding to Human PIWIL4. Recombinant protein control fragment (Product #RP-97318). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion