missing translation for 'onlineSavingsMsg'
Learn More

PIWIL2 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30733478 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733478 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733478 Supplier Novus Biologicals Supplier No. AF6558Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PIWIL2 Polyclonal antibody specifically detects PIWIL2 in Human,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIWIL2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2
Host Species Sheep
Immunogen E. coli-derived recombinant human PIWIL2, Lys216-Asp379, Accession # Q8TC59
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 55124
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.