missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93688-0.02ml
This item is not returnable.
View return policy
Description
PIWIL2 Polyclonal antibody specifically detects PIWIL2 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| PIWIL2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:3000 | |
| Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 398-652 of mouse PIWIL2 (Q8CDG1). QNKEHFQDECSKLLVGSIVITRYNNRTYRIDDVDWNKTPKDSFVMSDGKEITFLEYYSKNYGITVKEDDQPLLIHRPSERQNNHGMLLKGEILLLPELSFMTGIPEKMKKDFRAMKDLTQQINLSPKQHHGALECLLQRISQNETASNELTRWGLSLHKDVHKIEGRLLPMERINLRNTSFVTSEDLNWVKEVTRDASILTIPMHFWALFYPKRAMDQARELVNMLEKIAGPIGMRISPPAWVELKDDRIETYIR | |
| 0.02 mL | |
| Stem Cells | |
| 55124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction