missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIWIL1 Recombinant Rabbit Monoclonal Antibody (24GB1545)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557606
This item is not returnable.
View return policy
Description
PIWIL1 Recombinant Monoclonal Antibody for Western Blot, Flow
Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
Specifications
| PIWIL1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PIWIL1 | |
| CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 363912, 57749, 9271 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1545 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96J94, Q9JMB7 | |
| PIWIL1 | |
| A synthesized peptide derived from human PIWIL1 (aa 754-800). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction