missing translation for 'onlineSavingsMsg'
Learn More

PIWIL1/HIWI Antibody [CoraFluor™ 1], Novus Biologicals™

Product Code. 30732264 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732264 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732264 Supplier Novus Biologicals Supplier No. AF6548CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PIWIL1/HIWI Polyclonal antibody specifically detects PIWIL1/HIWI in Human,Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIWIL1/HIWI
Applications Immunohistochemistry
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias HIWIpiwi homolog, MIWI, PIWI, piwi (Drosophila)-like 1, piwi-like 1 (Drosophila), piwi-like protein 1
Host Species Goat
Immunogen E. coli-derived recombinant human PIWIL1/HIWI, Arg82-Thr290, Accession # Q96J94
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9271
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.