missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pivalic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2-Dimethylpropionic acid |
| CAS | 75-98-9 |
| Molecular Formula | C5 H10 O2 |
| Quantity | 100 g |
| Synonym | Propanoic acid','2,2-dimethyl- (9CI','ACI), 2,2-Dimethylpropanoic acid (ACI), Pivalic acid (7CI','8CI), 2,2,2-Trimethylacetic acid, 2,2-Dimethylpropionic acid, Neopentanoic acid, Neovaleric acid, NSC 65449, tert-Pentanoic acid, Trimethylacetic acid, Trimethylmethanecarboxylic acid, Versatic 5 acid, Versatic Acid 5, α,α-Dimethylpropionic acid |
| InChI Formula | InChI=1S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7) |
| SMILES | CC(C)(C)C(=O)O |
| IUPAC Name | 2,2-dimethylpropanoic acid |
| Molecular Weight (g/mol) | 102.13 |
| Formula Weight | 102.0681 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?