missing translation for 'onlineSavingsMsg'
Learn More

Pivaldehyde, 96%, AcroSeal™

Product Code. 10142514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10142514 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142514 Supplier Thermo Scientific Acros Supplier No. 427201000
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 630-19-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 630-19-3
Melting Point 6.0°C
Color Colorless
Density 0.7750g/mL
Boiling Point 75.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H10O
Linear Formula (CH3)3CCHO
Refractive Index 1.3775 to 1.3795
MDL Number MFCD00006962
Quantity 100 mL
Merck Index 15, 7624
Specific Gravity 0.775
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
Solubility Information Solubility in water: negligibly soluble.
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
SMILES CC(C)(C)C=O
IUPAC Name 2,2-dimethylpropanal
Molecular Weight (g/mol) 86.13
PubChem CID 12417
Formula Weight 86.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Pivaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pivaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.