Learn More
PITX2 Mouse anti-Human, Unconjugated, Clone: 1E3B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67201-1-IG-20UL
 View more versions of this product
Product Code. 16809715
  • £135.00 / 20µL
Description
The PITX2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PITX2. This antibody recognizes human, mouse, rat antigen. The PITX2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PITX2
Monoclonal
800 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1E3B1
Unconjugated
20 μL
5308
IgG1