missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33337-100ul
This item is not returnable.
View return policy
Description
PITX2 Monoclonal antibody specifically detects PITX2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| PITX2 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| all1-responsive gene 1, ALL1-responsive protein ARP1, ARP1MGC20144, Homeobox protein PITX2, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, paired-like homeodomain 2, Paired-like homeodomain transcription factor 2MGC111022, pituitary homeo box 2, pituitary homeobox 2, PTX2, RGSRIEG1Brx1, RIEG, RIEG bicoid-related homeobox transcription factor, rieg bicoid-related homeobox transcription factor 1, RS, solurshin | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PITX2 (NP_700475.1).,, Sequence:, METNCRKLVSACVQLGVQPAAVECLFSKDSEIKKVEFTDSPESRKEAASSKFFPRQHPGANEKDKSQQGKNEDVGAEDPSKKKRQRRQRTHFTSQQLQEL | |
| 100 μL | |
| Stem Cell Markers | |
| 5308 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction