missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PITX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552600
This item is not returnable.
View return policy
Description
Immunogen sequence: SFTFFNSMSP LSSQSMFSAP SSISSMTMPS SMGPGAVPGM PNSGLNNINN LTGSSLNSAM SPGACPYGTP ASPYSVYRDT CNSSLASLRL KSKQHSS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
PITX1 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family are involved in organ development and left-right asymmetry. This protein acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin.
Specifications
| PITX1 | |
| Polyclonal | |
| Unconjugated | |
| PITX1 | |
| BFT; CCF; hindlimb expressed homeobox protein backfoot; hindlimb-expressed homeobox protein backfoot; homeobox protein PITX1; Homeobox protein P-OTX; LBNBG; OTX; paired like homeodomain 1; paired-like homeodomain 1; paired-like homeodomain transcription factor 1; pituitary homeo box 1; pituitary homeobox 1; Pituitary OTX-related factor; Pitx1; Potx; P-OTX; PTX1; Unknown (protein for MGC:136976); zgc:136976 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78337 | |
| PITX1 | |
| Recombinant protein corresponding to Human PITX1. Recombinant protein control fragment (Product #RP-89293). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction