missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PITRM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531558
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (84%), Rat (81%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an ATP-dependent metalloprotease that degrades post-cleavage mitochondrial transit peptides. The encoded protein binds zinc and can also degrade amyloid beta A4 protein, suggesting a possible role in Alzheimer's disease.
Specifications
| PITRM1 | |
| Polyclonal | |
| Unconjugated | |
| PITRM1 | |
| 2310012C15Rik; AA410010; hPreP; KIAA1104; Metalloprotease 1; metalloprotease 1 (pitrilysin family); mKIAA1104; MP1; MP-1; Ntup1; nuclear transplantation upregulated protein 1; pitrilysin metallepetidase 1; pitrilysin metallopeptidase 1; pitrilysin metalloprotease 1; pitrilysin metalloproteinase 1; Pitrm1; PREP; PreP peptidasome; presequence protease, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10531 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q5JRX3 | |
| PITRM1 | |
| Recombinant protein encompassing a sequence within the center region of human PITRM1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction