missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PITPNM3 Polyclonal Antibody

Product Code. 15742521 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15742521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15742521 Supplier Invitrogen™ Supplier No. PA554589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RFPLGDGQSL LLADALHTHS PLFLEGSSRD SPPLLDAPAS PPQASRFQRP GRRMSEGSSH SESSESSDSM APVGASRITA KWWGSKRIDY ALYCPDV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

This gene encodes a member of a family of membrane-associated phosphatidylinositol transfer domain-containing proteins. The calcium-binding protein has phosphatidylinositol transfer activity and interacts with the protein tyrosine kinase PTK2B. The protein is homologous to a Drosophila protein that is implicated in the visual transduction pathway in flies. Mutations in this gene result in autosomal dominant cone dystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITPNM3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PITPNM3
Gene Accession No. Q9BZ71
Gene Alias A330068P14Rik; ACKR6; AI848332; atypical chemokine receptor 6; cone rod dystrophy 5; CORD5; Gm880; Membrane-associated phosphatidylinositol transfer protein 3; MGC157740; MGC157741; NIR1; NIR-1; phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PITPNM family member 3; Pitpnm3; Pyk2 N-terminal domain-interacting receptor 1; RDGBA3; retinal degeneration B alpha 3
Gene Symbols PITPNM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PITPNM3. Recombinant protein control fragment (Product #RP-93182).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83394
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.