missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PITPNM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566530

Product Code. 15989224

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SRRGSMNNELL SPEFGPVRDP LADGVEGLGR GSPEPSALPP QRIPSDMASP EPEGSQNSLQ AA Highest antigen sequence identity to the following orthologs - mouse 83%, rat 83%.

Regulates RHOA activity, and plays a role in cytoskeleton remodeling. Necessary for normal completion of cytokinesis. Plays a role in maintaining normal diacylglycerol levels in the Golgi apparatus. Binds phosphatidyl inositol phosphates (in vitro). May catalyze the transfer of phosphatidylinositol and phosphatidylcholine between membranes. Necessary for maintaining the normal structure of the endoplasmic reticulum and the Golgi apparatus. Required for protein export from the endoplasmic reticulum and the Golgi. Binds calcium ions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PITPNM1
Polyclonal
Unconjugated
PITPNM1
Dres9; drosophila retinal degeneration B homolog; Drosophila retinal degeneration B homolog 1; FLJ44997; membrane-associated phosphatidylinositol transfer protein 1; Mpt1; Mpt-1; NIR2; Nir-2; phosphatidylinositol membrane-associated; phosphatidylinositol transfer protein membrane associated 1; phosphatidylinositol transfer protein, membrane associated 1; phosphatidylinositol transfer protein, membrane-associated 1; Pitpnm; PITPnm 1; PITPNM1; Pyk2 N-terminal domain-interacting receptor 2; R75447; Rd9; RdgB; RDGB1; RDGBA; RDGBA1; retinal degeneration 9; retinal degeneration B alpha 1
Rabbit
Antigen affinity chromatography
RUO
9600
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O00562
PITPNM1
Recombinant Human PITPNM1. Recombinant protein control fragment (Product #RP-107193).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.