missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PITPNB Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531358
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431 , Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (85%), Chicken (92%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a cytoplasmic protein that catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes. This transfer activity is required for COPI complex-mediated retrograde transport from the Golgi apparatus to the endoplasmic reticulum. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| PITPNB | |
| Polyclonal | |
| Unconjugated | |
| PITPNB | |
| phosphatidylinositol transfer protein beta; phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; Pitp beta; PI-TP beta; PI-TP-beta; Pitpnb; ptdIns transfer protein beta; PtdInsTP; ptdInsTP beta; VIB1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23760 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P48739 | |
| PITPNB | |
| Recombinant protein encompassing a sequence within the center region of human PITPNB. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction