missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITPNB Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18403R
This item is not returnable.
View return policy
Description
PITPNB Polyclonal antibody specifically detects PITPNB in Human, Mouse samples. It is validated for Western Blot
Specifications
| PITPNB | |
| Polyclonal | |
| Western Blot | |
| phosphatidylinositol transfer protein beta isoform, phosphatidylinositol transfer protein, beta, phosphotidylinositol transfer protein, beta, PI-TP-beta, PtdIns transfer protein beta, PtdInsTP, PtdInsTP beta, VIB1B | |
| Recognizes region between residues Between Between 221 and 271.. | |
| 0.1 mL | |
| metabolism | |
| 23760 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction