missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PITPN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104010
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PITPNA.
This gene encodes a member of a family of lipid-binding proteins that transfer molecules of phosphatidylinositol or phosphatidylcholine between membrane surfaces. The protein is implicated in phospholipase C signaling and in the production of phosphatidylinositol 3,4,5-trisphosphate (PIP3) by phosphoinositide-3-kinase.
Specifications
| PITPN | |
| Polyclonal | |
| Unconjugated | |
| PITPNA | |
| epididymis secretory protein Li 36; HEL-S-36; phosphatidylinositol transfer protein alpha; phosphatidylinositol transfer protein alpha isoform; phosphatidylinositol transfer protein, alpha; Pitp alpha; PI-TPalpha; PI-TP-alpha; PITPN; Pitpna; PITPNB; ptdIns transfer protein alpha; ptdInsTP alpha; vb; VIB1A; vibrator | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18738, 29525, 5306 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P16446, P53810, Q00169 | |
| PITPNA | |
| A synthesized peptide derived from human PITPNA(Accession Q00169), corresponding to amino acid residues L150-F200. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction