missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PITPN Antibody (4G10), Novus Biologicals™

Product Code. 18344169 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344169 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344169 Supplier Novus Biologicals™ Supplier No. H00005306M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PITPN Monoclonal antibody specifically detects PITPN in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITPN
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 4G10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006215
Gene Alias MGC99649, phosphatidylinositol transfer protein alpha isoform, phosphatidylinositol transfer protein, alpha, PI-TPalpha, PI-TP-alpha, PITPNphosphotidylinositol transfer protein, PtdIns transfer protein alpha, PtdInsTP alpha, VIB1A
Host Species Mouse
Immunogen PITPNA (NP_006215.1, 171 a.a. ∽ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GPLGPNWKQELVNQKDCPYMCAYKLVTVKFKWWGLQNKVENFIHKQERRLFTNFHRQLFCWLDKWVDLTMDDIRRMEEETKRQLDEMRQKDPVKGMTADD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 5306
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.