missing translation for 'onlineSavingsMsg'
Learn More

Pitolisant, TRC

Product Code. 30415283
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415283 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415283 Supplier Toronto Research Chemicals Supplier No. TRCP5365051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pitolisant
CAS 362665-56-3
Molecular Formula C17 H26 Cl N O
Quantity 1 g
Synonym Piperidine','1-[3-[3-(4-chlorophenyl)propoxy]propyl]-, 1-[3-[3-(4-Chlorophenyl)propoxy]propyl]piperidine, 3-(4-Chlorophenyl)propyl 3-piperidinopropyl ether, Pitolisant
InChI Formula InChI=1S/C17H26ClNO/c18-17-9-7-16(8-10-17)6-4-14-20-15-5-13-19-11-2-1-3-12-19/h7-10H,1-6,11-15H2
SMILES Clc1ccc(CCCOCCCN2CCCCC2)cc1
IUPAC Name 1-[3-[3-(4-chlorophenyl)propoxy]propyl]piperidine
Molecular Weight (g/mol) 295.847
Formula Weight 295.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.