missing translation for 'onlineSavingsMsg'
Learn More

Pitolisant oxalate, MedChemExpress

Product Code. 16415018
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16415018

Brand: Medchem Express HY12199A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Pitolisant oxalate is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

Pitolisant oxalate is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

Specifications

Chemical Name or Material Pitolisant oxalate
Physical Form Solid
CAS 362665-57-4
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H28ClNO5
Synonym Tiprolisant oxalate
Solubility Information DMSO : 50 mg/mL (129.57 mM; Need ultrasonic)
SMILES ClC1=CC=C(CCCOCCCN2CCCCC2)C=C1.O=C(O)C(O)=O
Molecular Weight (g/mol) 385.88
Formula Weight 385.88
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.