missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pitolisant, MedChemExpress
Description
Pitolisant is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).
Specifications
Specifications
| Chemical Name or Material | Pitolisant |
| Physical Form | Liquid |
| CAS | 362665-56-3 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H26ClNO |
| Synonym | Tiprolisant |
| Solubility Information | DMSO : 100 mg/mL (338.01 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C(CCCOCCCN2CCCCC2)C=C1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?