missing translation for 'onlineSavingsMsg'
Learn More

Pitolisant, MedChemExpress

Product Code. 16401257
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16401257

Brand: Medchem Express HY12199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pitolisant is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

Pitolisant is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

Specifications

Chemical Name or Material Pitolisant
Physical Form Liquid
CAS 362665-56-3
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H26ClNO
Synonym Tiprolisant
Solubility Information DMSO : 100 mg/mL (338.01 mM; Need ultrasonic)
SMILES ClC1=CC=C(CCCOCCCN2CCCCC2)C=C1
Molecular Weight (g/mol) 295.85
Formula Weight 295.85
Percent Purity 97.22%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.