missing translation for 'onlineSavingsMsg'
Learn More

Pitolisant hydrochloride, MedChemExpress

Product Code. 16425018
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16425018

Brand: Medchem Express HY12199B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

Specifications

Chemical Name or Material Pitolisant hydrochloride
Physical Form Solid
CAS 903576-44-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H27Cl2NO
For Use With (Application) Neuroscience-Neuromodulation
Synonym Ciproxidine BF 2649
Solubility Information H2O : 100 mg/mL (300.92 mM; Need ultrasonic) ∣DMSO : ≥ 43 mg/mL (129.40 mM)
SMILES ClC(C=C1)=CC=C1CCCOCCCN2CCCCC2.Cl
Molecular Weight (g/mol) 332.31
Formula Weight 332.31
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.