missing translation for 'onlineSavingsMsg'
Learn More

Pit1 Antibody, Novus Biologicals™

Product Code. 18362344 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362344 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362344 Supplier Novus Biologicals Supplier No. NBP303868

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pit1 Polyclonal specifically detects Pit1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pit1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CPHD1, GHF1, GHF-1pituitary-specific positive transcription factor 1, Growth hormone factor 1, PIT-1, PIT1pituitary-specific transcription factor 1, POU class 1 homeobox 1, POU domain class 1, transcription factor 1, POU domain, class 1, transcription factor 1, POU1F1a
Host Species Rabbit
Immunogen Recombinant protein of human POU1F1.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5449
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.