missing translation for 'onlineSavingsMsg'
Learn More

PIST Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18406893 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406893 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406893 Supplier Novus Biologicals Supplier No. NBP177242AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIST Polyclonal antibody specifically detects PIST in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIST
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CALPDZ protein interacting specifically with TC10, CFTR-associated ligand, dJ94G16.2, FIGdJ94G16.2 PIST, Fused in glioblastoma, golgi-associated PDZ and coiled-coil motif containing, Golgi-associated PDZ and coiled-coil motif-containing protein, GOPC1, PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10, PISTGolgi associated PDZ and coiled-coil motif containing protein
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human PIST. The immunogen is located within amino acids 360 - 410 of PIST. Amino Acid Squence: APEVDSDDENVEYEDE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57120
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.