Learn More
PISD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16401-1-AP-20UL
 View more versions of this product
Product Code. 16886014
  • £135.00 / 20µL
Description
This antibody is specific to PISD. The sequence corresponds to the beta isoform.
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
Specifications
Specifications
PISD
Polyclonal
Unconjugated
PISD
DJ858B16, PISD, PSDC, PSSC
Rabbit
Antigen Affinity Chromatography
RUO
23761, 320951, 681361
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZAW2, Q8BSF4, Q9UG56
Pisd
PISD Fusion Protein Ag9499
20 μL
Primary
Mouse, Human, Rat