missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PISD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16401-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to PISD. The sequence corresponds to the beta isoform.
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.Specifications
| PISD | |
| Polyclonal | |
| Unconjugated | |
| PISD | |
| DJ858B16, PISD, PSDC, PSSC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23761, 320951, 681361 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZAW2, Q8BSF4, Q9UG56 | |
| Pisd | |
| PISD Fusion Protein Ag9499 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction