missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PISD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556634
This item is not returnable.
View return policy
Description
Immunogen sequence: YKSVPTRLLS RAWGRLNQVE LPHWLRRPVY SLYIWTFGVN MKEAAVEDLH HYRNLSEFFR RKLKPQARPV CGLHSISPSD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
Specifications
| PISD | |
| Polyclonal | |
| Unconjugated | |
| PISD | |
| 9030221M09Rik; DJ858B16; dJ858B16.2; DKFZp566G2246; phosphatidylserine decarboxylase; Phosphatidylserine decarboxylase alpha chain; Phosphatidylserine decarboxylase beta chain; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; Pisd; PSD; PSDC; PSSC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23761 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UG56 | |
| PISD | |
| Recombinant protein corresponding to Human PISD. Recombinant protein control fragment (Product #RP-95350). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction