missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIRT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144340
This item is not returnable.
View return policy
Description
PIRT Polyclonal Antibody for ICC/IF, IHC (P), ELISA
PIRT is a key component of the TRPV1 complex that positively regulates TRPV1 activity. It binds various phosphoinositide, including phosphatidylinositol 4, 5-bisphosphate (PIP2), but not phosphatidylinositol (PI). PIRT is a membrane protein with MW 15-25 kDa.
Specifications
| PIRT | |
| Polyclonal | |
| Unconjugated | |
| PIRT | |
| A530088H08Rik; phosphoinositide interacting protein; phosphoinositide interacting regulator of transient receptor potential channels; phosphoinositide-interacting protein; phosphoinositide-interacting regulator of transient receptor potential channels; phosphoinositide-interacting regulator of TRPV1; Pirt; RGD1565284 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 644139 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.79 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P0C851 | |
| PIRT | |
| Recombinant Human Phosphoinositide-interacting protein (1-55AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction