missing translation for 'onlineSavingsMsg'
Learn More

Piroxicam, TRC

Product Code. 30409504
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409504 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409504 Supplier Toronto Research Chemicals Supplier No. TRCP5100005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Piroxicam
CAS 36322-90-4
Purity Grade Notes HPLC
Molecular Formula C15 H13 N3 O4 S
Quantity 5 g
Synonym 4-Hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, Artroxicam, Baxo, Bruxicam, CHF 1251, CP 16171, Caliment, Dispercam, Doblexan, Dolibid, Erazon, Felden gel, Feldene, Feldene Melt, Feldene Zydis, Feldoral, Feledene, Feloxin, Felvin 20, Felxicam, Flogobene, Flogosan, Geldene, Improntal, Larapam, Laxidene, Maxvin, Murupe, NSC 666076, Neoxicam, Nesprex-DT, Nkoyo piroxicam, Pirkam, Piroflex, Piroftal, Piroxicam, Piroxidine, Piroxitas-DT, Pixicam, Pyroxycam, Reudene, Rheugesic, Riacen, Roxicam, Roxiden, Sasulen, Solocalm, Uphaxicam, Zunden
InChI Formula InChI=1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)
SMILES CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ccccn3
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1λ^{6},2-benzothiazine-3-carboxamide
Molecular Weight (g/mol) 331.35
Formula Weight 331.0627
Percent Purity >95
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.