Learn More
Piroxicam, Thermo Scientific™
£37.81 - £221.99
Chemical Identifiers
| CAS | 36322-90-4 |
|---|---|
| Molecular Formula | C15H13N3O4S |
| Molecular Weight (g/mol) | 331.346 |
| MDL Number | MFCD00057317 |
| InChI Key | QYSPLQLAKJAUJT-UHFFFAOYSA-N |
| Synonym | piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment |
| PubChem CID | 54676228 |
| ChEBI | CHEBI:8249 |
| IUPAC Name | 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide |
| SMILES | CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15435499
|
Thermo Scientific Alfa Aesar
J63239.03 |
1g |
£38.60 £37.81 / 1g Save £0.79 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15445499
|
Thermo Scientific Alfa Aesar
J63239.06 |
5g |
£122.00 £119.00 / 5g Save £3.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15455499
|
Thermo Scientific Alfa Aesar
J63239.09 |
£227.00 £221.99 / 10g Save £5.01 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsPiroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.
Solubility
Soluble in water, ethanol, chloroform, ethyl acetate.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 36322-90-4 | |
| 331.346 | |
| QYSPLQLAKJAUJT-UHFFFAOYSA-N | |
| 54676228 | |
| 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide |
| C15H13N3O4S | |
| MFCD00057317 | |
| piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment | |
| CHEBI:8249 | |
| CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3 |
Specifications
| 36322-90-4 | |
| MFCD00057317 | |
| 14,7506 | |
| Soluble in water,ethanol,chloroform,ethyl acetate. | |
| CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3 | |
| 331.346 | |
| CHEBI:8249 | |
| Piroxicam |
| C15H13N3O4S | |
| UN2811 | |
| piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment | |
| QYSPLQLAKJAUJT-UHFFFAOYSA-N | |
| 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide | |
| 54676228 | |
| 331.35 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 252-974-3
missing translation for 'rtecsNumber' : DL0705000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- Piroxicam
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
- MIXTURE LIST-Contains : Piroxicam
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.