missing translation for 'onlineSavingsMsg'
Learn More

Piroxicam 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30458121
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30458121

Brand: LoGiCal LGCAMP0084.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Piroxicam 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Piroxicam
CAS 36322-90-4
Analyte or Component Names Piroxicam
Shipping Condition Room Temperature
Molecular Formula C15 H13 N3 O4 S
Certifications/Compliance ISO 17034
Synonym 2 H-1,2-Benzothiazine-3-carboxamide, 4-hydroxy-2-methyl-N-2-pyridinyl-, 1,1-dioxide, 4-Hydroxy-2-methyl-N-(2-pyridyl)-2 H-1,2-benzothiazine-3-carboxamide 1,1-dioxide, Artroxicam, Baxo, Bruxicam, CHF 1251, CP 16171, Caliment, Dispercam, Doblexan, Dolibid, Erazon, Felden gel, Feldene, Feldene Melt, Feloxin, Felvin 20, Felxicam, Flogobene, Geldene, Improntal, Larapam, Laxidene, Maxvin, NSC 666076, Neoxicam, Nkoyo piroxicam, Pirkam, Piroflex, Piroftal, Piroxicam, Piroxidine, Pixicam, Pyroxycam, Reudene, Rheugesic, Riacen, Roxicam, Roxiden, Sasulen, Solocalm, Uphaxicam, Zunden, Zydis, Feldene Zydis, Feldoral, Feledene, Flogosan, Murupe, Nesprex-DT, Piroxitas-DT
InChI Formula InChI=1 S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19 H,1H3,(H,16,17,20)
SMILES CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ccccn3
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1λ^{6},2-benzothiazine-3-carboxamide
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 331.35
Quantity 1 mL
Formula Weight 331.0627 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.