missing translation for 'onlineSavingsMsg'
Learn More

Piroctone olamine, Thermo Scientific™

Product Code. 17700639 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17700639

Brand: Thermo Scientific Acros 468820010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Alpha Vector PIROCTONEOLAMINE
Chemical Name or Material Piroctone olamine
CAS 68890-66-4
Melting Point 130°C to 138°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C14 H23 N O2 . C2 H7 N O
MDL Number 01690792
Quantity 1 g
Merck Index XIII 7585
Molecular Weight (g/mol) 298.46
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.