missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pirlimycin Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA175351

Product Code. 11574370

  • £466.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Working Concentration: 2.5 μg/mL. Sensitivity: 10 ng/mL Pirlimycin produces 29% inhibition in a competitive ELISA employing Pirlimycin polyclonal antibody.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pirlimycin
Polyclonal
PBS with 0.09% sodium azide; pH 7.2
Pirlimycin (N) -BTG.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA
Unconjugated
Sheep
Purified
RUO
Chemical
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.