missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pirin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529777
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, mouse liver. Predicted reactivity: Mouse (95%), Rat (96%), Rhesus Monkey (98%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the cupin superfamily. The encoded protein is an Fe(II)-containing nuclear protein expressed in all tissues of the body and concentrated within dot-like subnuclear structures. Interactions with nuclear factor I/CCAAT box transcription factor as well as B cell lymphoma 3-encoded oncoprotein suggest the encoded protein may act as a transcriptional cofactor and be involved in the regulation of DNA transcription and replication. Alternatively spliced transcript variants have been described.
Specifications
| Pirin | |
| Polyclonal | |
| Unconjugated | |
| PIR | |
| 2310042L19Rik; iron-binding nuclear protein; PIR; Pirin; pirin (iron-binding nuclear protein); pirin like; pirinin; Pirnl; Probable quercetin 2,3-dioxygenase PIR; probable quercetinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 363465, 69656, 8544 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.96 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O00625, Q5M827, Q9D711 | |
| PIR | |
| Recombinant protein encompassing a sequence within the center region of human Pirin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction