missing translation for 'onlineSavingsMsg'
Learn More

Piribedil, MedChemExpress

Product Code. p-200100398
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16485298

Brand: Medchem Express HY12707

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Piribedil is a potent and orally active dopamine D2 and dopamine D3 agonist. Piribedil is also a α2-adrenoceptors antagonist. Piribedil can inhibit MLL1 methyltransferase activity (EC50: 0.18 μM). Piribedil has the potential for the research of parkinson's disease, circulatory disorders, cancers.

Piribedil is a potent and orally active dopamine D2 and dopamine D3 agonist. Piribedil is also a α2-adrenoceptors antagonist. Piribedil can inhibit MLL1 methyltransferase activity (EC50: 0.18 μM). Piribedil has the potential for the research of parkinson's disease, circulatory disorders, cancers.

Specifications

Chemical Name or Material Piribedil
CAS 3605-01-4
Color White
Purity Grade Notes Research
Molecular Formula C16H18N4O2
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (111.72 mM; Need ultrasonic)
SMILES C1(N2CCN(CC3=CC=C(OCO4)C4=C3)CC2)=NC=CC=N1
Molecular Weight (g/mol) 298.34
Formula Weight 298.34
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.