missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pirh2 Recombinant Rabbit Monoclonal Antibody (K01_3K28)

Product Code. 30828590
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828590 100 μL
30827388 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828590 Supplier Invitrogen™ Supplier No. MA564344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Pirh2 Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene has ubiquitin-protein ligase activity. This protein binds with p53 and promotes the ubiquitin-mediated proteosomal degradation of p53. This gene is oncogenic because loss of p53 function contributes directly to malignant tumor development. Transcription of this gene is regulated by p53. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pirh2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3K28
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene RCHY1
Gene Accession No. Q96PM5
Gene Alias Androgen receptor N-terminal-interacting protein; androgen-receptor N-terminal-interacting protein; ARNIP; Chimp; CH-rich interacting match with PLAG1; CH-rich-interacting match with PLAG1; DKFZp586C1620; E3 ubiquitin-protein ligase Pirh2; h PRO1996; hPirh2; p53-induced protein with a RING-H2 domain; p53-induced RING-H2 protein; PIRH2; PIRH2E; PIRH2F; PRO1996; RCHY1; ring finger and CHY zinc finger domain containing 1; ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase; RING finger and CHY zinc finger domain-containing protein 1; RING finger protein 199; RING-type E3 ubiquitin transferase RCHY1; RNF199; ZCHY; Zfp363; Zinc finger protein 363; zinc finger, CHY-type; Znf363
Gene Symbols RCHY1
Host Species Rabbit
Immunogen Recombinant protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25898, 289508
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.