missing translation for 'onlineSavingsMsg'
Learn More

Pirh2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18357204
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357204 25 μg 25µL
18309372 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18357204 Supplier Novus Biologicals Supplier No. NBP31729125UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pirh2 Polyclonal antibody specifically detects Pirh2 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pirh2
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias Androgen receptor N-terminal-interacting protein, androgen-receptor N-terminal-interacting protein, ARNIPRING finger protein 199, CHIMPRNF199DKFZp586C1620, CH-rich interacting match with PLAG1, CH-rich-interacting match with PLAG1, E3 ubiquitin-protein ligase Pirh2, EC 6.3.2.-, hARNIP, hPirh2, PIRH2, PRO1996, ring finger and CHY zinc finger domain containing 1, RING finger and CHY zinc finger domain-containing protein 1, Zinc finger protein 363p53-induced RING-H2 protein, ZNF363
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: TAREDGASGQERGQRGCEHYDRGCLLKAPCCDKLYTCRLCHDNNEDHQLDRFKVKEVQCINCEKIQHAQQTCEECSTLFGEYY
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, p53 Pathway, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25898
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.