missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pirh2 Rabbit anti-Human, Mouse, Clone: 4Q5H9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15743-100UL
This item is not returnable.
View return policy
Description
Pirh2 Monoclonal antibody specifically detects Pirh2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Pirh2 | |
| Monoclonal | |
| Unconjugated | |
| Androgen receptor N-terminal-interacting protein, androgen-receptor N-terminal-interacting protein, ARNIPRING finger protein 199, CHIMPRNF199DKFZp586C1620, CH-rich interacting match with PLAG1, CH-rich-interacting match with PLAG1, E3 ubiquitin-protein ligase Pirh2, EC 6.3.2.-, hARNIP, hPirh2, PIRH2, PRO1996, ring finger and CHY zinc finger domain containing 1, RING finger and CHY zinc finger domain-containing protein 1, Zinc finger protein 363p53-induced RING-H2 protein, ZNF363 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-261 of human Pirh2 (Q96PM5). | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer, p53 Pathway, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| 25898 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4Q5H9 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction