missing translation for 'onlineSavingsMsg'
Learn More

Pirh2 Antibody (5D12), Novus Biologicals™

Product Code. 18393119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393119 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393119 Supplier Novus Biologicals Supplier No. H00025898M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pirh2 Monoclonal antibody specifically detects Pirh2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pirh2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5D12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_056251
Gene Alias Androgen receptor N-terminal-interacting protein, androgen-receptor N-terminal-interacting protein, ARNIPRING finger protein 199, CHIMPRNF199DKFZp586C1620, CH-rich interacting match with PLAG1, CH-rich-interacting match with PLAG1, E3 ubiquitin-protein ligase Pirh2, EC 6.3.2.-, hARNIP, hPirh2, PIRH2, PRO1996, ring finger and CHY zinc finger domain containing 1, RING finger and CHY zinc finger domain-containing protein 1, Zinc finger protein 363p53-induced RING-H2 protein, ZNF363
Host Species Mouse
Immunogen RCHY1 (NP_056251, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAATAREDGASGQERGQRGCEHYDRGCLLKAPCCDKLYTCRLCHDNNEDHQLDRFKVKEVQCINCEKIQHAQQTCEECSTLFGEYYCDICHLFDKDKKQY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, p53 Pathway, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 25898
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.