missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine dihydrochloride, Tocris Bioscience™

Product Code. 18034482
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
18034482 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 18034482

Brand: Tocris Bioscience 1071/100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective M1 antagonist

M1 muscarinic receptor selective antagonist. Inverse agonist activity reported.
TRUSTED_SUSTAINABILITY

Specifications

CAS 29868-97-1
Molecular Formula C19H23Cl2N5O2
Quantity 100 mg
Synonym pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
InChI Key FFNMBRCFFADNAO-UHFFFAOYSA-N
SMILES CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl
IUPAC Name 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
Molecular Weight (g/mol) 424.326
PubChem CID 71405
ChEBI CHEBI:32014
Formula Weight 424.33
Percent Purity >98%
Chemical Name or Material Pirenzepine dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.